Synonym: Ammonium vanadate(V)
CAS Number 7803-55-6
Linear Formula NH4VO3
Molecular Weight 116.98
EC Number 232-261-3
MDL number MFCD00011430
PubChem Substance ID 24852206
Popular Documents: Specification Sheet (PDF)
| assay | 99% |
| density | 2.32 g/cm3 at 25 °C(lit.) |
1 kg in poly bottle
250 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?