• USA Home
  • 207306 - Molybdenum atomic absorption standard solution

EMAIL THIS PAGE TO A FRIEND
207306 Sigma-Aldrich

Molybdenum atomic absorption standard solution

analytical standard, 1,000 μg/mL Mo in H2O (Exact Mo concentration on label)

Purchase

Properties

Related Categories AA Standard Solutions, AAS, Additional Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
concentration   1,000 μg/mL Mo in H2O (Exact Mo concentration on label)
application(s)   ICP/AAS: suitable
density   1.005 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

67210 Molybdenum Standard for AAS, TraceCERT®, 1000 mg/L Mo in hydrochloric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?