EMAIL THIS PAGE TO A FRIEND
20740 Sigma-Aldrich

γ-Butyrolactone

puriss., ≥99.0% (GC)

Synonym: γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

  • CAS Number 96-48-0

  • Empirical Formula (Hill Notation) C4H6O2

  • Molecular Weight 86.09

  •  Beilstein Registry Number 105248

  •  EC Number 202-509-5

  •  MDL number MFCD00005386

  •  PubChem Substance ID 329752019

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Cell Biology, Chemical Synthesis, Lactones,
vapor density   3 (vs air)
vapor pressure   1.5 mmHg ( 20 °C)
InChI Key   YEJRWHAVMIAJKC-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   851 °F
expl. lim.   16 %
refractive index   n20/D 1.436(lit.)
  n20/D 1.437
bp   204-205 °C(lit.)
mp   −45 °C(lit.)
density   1.12 g/mL at 25 °C(lit.)

Description

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
LU3500000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B103608 γ-Butyrolactone, ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?