• USA Home
  • 207578 - Thorium Standard for AAS

EMAIL THIS PAGE TO A FRIEND
207578 Sigma-Aldrich

Thorium Standard for AAS

1000 μg/mL Th in 5% HNO3 (Exact Th concentration on label)

Purchase

Properties

Related Categories AA Standard Solutions, AAS, Additional Standards, Analytical Standards, Analytical/Chromatography,
concentration   1000 μg/mL Th in 5% HNO3 (Exact Th concentration on label)
application(s)   ICP/AAS: suitable
density   1.03 g/mL at 25 °C
format   single component solution

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2031B 8 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?