CAS Number 7553-56-2
Empirical Formula (Hill Notation) I2
Molecular Weight 253.81
Beilstein Registry Number 3587194
EC Number 231-442-4
MDL number MFCD00011355
PubChem Substance ID 24852469
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, C-X Bond Formation (Halogen), Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| vapor density | 9 (vs air) |
| vapor pressure | 0.31 mmHg ( 25 °C) |
| 1 mmHg ( 38.7 °C) | |
| assay | ≥99.8% |
| form | solid |
| resistivity | 1.3E15 μΩ-cm |
1, 2.5 kg in glass bottle
12 kg in steel drum
5, 100, 500, 6×500 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?