Synonym: Dichlorozinc
CAS Number 7646-85-7
Linear Formula ZnCl2
Molecular Weight 136.30
EC Number 231-592-0
MDL number MFCD00011295
PubChem Substance ID 24852492
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, |
| grade | reagent grade |
| vapor pressure | 1 mmHg ( 428 °C) |
| assay | ≥98% |
| mp | 293 °C(lit.) |
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?