Synonym: Molecular Sieve UOP Type 3Å
CAS Number 308080-99-1
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
MDL number MFCD00147627
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Molecular Sieves, Research Essentials |
| form | beads |
| particle size | 8-12 mesh |
1 kg in poly bottle
5 kg in fiber drum
Customers Also Viewed
beads
rod, size 1/16 in.
beads, 4-8 mesh
pellets, 3.2 mm
pellets, 1.6 mm
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?