CAS Number 63231-69-6
Linear Formula Na86[AlO2)86(SiO2)106] · xH2O
MDL number MFCD00131613
| Related Categories | Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Molecular Sieves, Research Essentials |
| form | beads |
| particle size | 8-12 mesh |
1 kg in poly bottle
5 kg in fiber drum
Customers Also Viewed
beads, 4-8 mesh
pellets, 3.2 mm
pellets, 1.6 mm
beads, 8-12 mesh
beads, 8-12 mesh
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?