EMAIL THIS PAGE TO A FRIEND
20890 Sigma-Aldrich

Cadmium

purum p.a., for metal reduction, 99.99%, granular

  • CAS Number 7440-43-9

  • Empirical Formula (Hill Notation) Cd

  • Molecular Weight 112.41

  •  Beilstein Registry Number 8137359

  •  EC Number 231-152-8

  •  MDL number MFCD00010914

  •  PubChem Substance ID 24852536

Purchase

Properties

Related Categories Cadmium, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science,
grade   for metal reduction
InChI Key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N
grade   purum p.a.
assay   99.99%
form   granular
resistivity   7.27 μΩ-cm, 22°C
impurities   iron, none detected
bp   765 °C(lit.)
mp   320.9 °C(lit.)
density   8.65 g/mL at 25 °C(lit.)
cation traces   Fe: ≤20 mg/kg

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Cross-Coupling Guide

Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2570 6.1/PG 1
WGK Germany 
3
RTECS 
EU9800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?