• USA Home
  • 20912 - Cadmium chloride hemi(pentahydrate)

EMAIL THIS PAGE TO A FRIEND
20912 Sigma-Aldrich

Cadmium chloride hemi(pentahydrate)

purum p.a., ≥98.0% (KT)

Purchase

Properties

Related Categories Cadmium, Cadmium Salts, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals,
vapor density   6.3 (vs air)
InChI Key   DZVRGWYMCGLNKJ-UHFFFAOYSA-J
grade   purum p.a.
assay   ≥98.0% (KT)
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Na: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2570 6.1/PG 3
WGK Germany 
3
RTECS 
EV0178000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?