209139 Sigma-Aldrich

Silver nitrate

ACS reagent, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Nitric acid silver(I) salt

Properties

Related Categories ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents,
grade   ACS reagent
vapor density   5.8 (vs air)
assay   ≥99.0%
form   solid
clarity of soln   passes test
impurities   Free acid, passes test
  ≤0.01% not HCl pptd.
mp   212 °C (dec.)(lit.)
anion traces   chloride (Cl-): ≤5 ppm
  sulfate (SO42-): ≤0.002%
cation traces   Cu: ≤2 ppm
  Fe: ≤2 ppm
  Pb: ≤0.001%

Description

Packaging

25, 100 g in amber poly

500 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Silver nitrate

BioXtra, >99% (titration)

Silver nitrate

BioReagent, suitable for plant cell culture, >99% (titration)

Silver nitrate

meets USP testing specifications

Silver nitrate

99.9999% trace metals basis

Silver nitrate

ReagentPlus®, ≥99.0% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?