EMAIL THIS PAGE TO A FRIEND
20967 Sigma-Aldrich

Cesium chloride

BioUltra, ≥99.0% (AT)

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Density Gradient, Salts of Alkali Metals
InChI Key   AIYUHDOJVYHVIT-UHFFFAOYSA-M
product line   BioUltra
assay   ≥99.0% (AT)
impurities   insoluble matter, passes filter test
  ≤0.001% total nitrogen (N)
loss   ≤0.1% loss on drying, 110 °C
pH   5.0-7.5 (25 °C, 3 M in H2O)
mp   645 °C(lit.)
solubility   H2O: soluble3 M at 20 °C, clear, colorless
anion traces   sulfate (SO42-): ≤20 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   3 M in H2O
UV absorption   λ: 260 nm Amax: 0.03
  λ: 280 nm Amax: 0.03

Description

Application

Used for the preparation of electrically conducting glasses. Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FK9625000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

20966 Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?