EMAIL THIS PAGE TO A FRIEND

210013 Sigma-Aldrich

Hydriodic acid

contains No stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis

Synonym: Hydriotic acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Alternative Energy, Chemical Synthesis, Halide Sources,
assay   99.99% trace metals basis
does not contain   stabilizer
concentration   57 wt. % in H2O
bp   127 °C(lit.)
mp   -50 °C
density   1.701 g/mL at 25 °C
storage temp.   2-8°C

Description

Packaging

50, 250 mL in glass bottle

Price and Availability

Customers Also Viewed

Hydriodic acid

57 wt. % in H2O, distilled, stabilized, 99.95%

Hydriodic acid

contains No stabilizer, ACS reagent, 55%

Hydriodic acid

contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%

Hydriodic acid

puriss. p.a., ≥67% (T)

Hydriodic acid

puriss. p.a., 55-60%, not stabilized, packed in coated, shock- and leak-protected glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1787 8 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Silylethyne-Substituted Pentacenes

Research into the use of organic semiconductors in field-effect transistors (FETs) began in earnest in the mid-1990s,1 after early exciting results from vapor-deposited small molecule semiconductors....
John E. Anthony*
Material Matters 2009, 4.3, 58.
Keywords: Applications, Capabilities, Crystallization, Deposition, Electronics, Electrophoretic display, Evaporation, Organic electronics, Semiconductor, Separation, Solar cells, Substitutions

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Relationships between circulating and intracellular thyroid hormones: physiological and clinical implications. Larsen PR, Silva JE, and Kaplan MM Endocr. Rev. 2(1), 87-102, (1981)

Loading...


Novel method for the determination of the methoxyl content in lignin by headspace gas chromatography. Li H, Chai XS, Liu M, et al. J. Agric. Food Chem. 60(21), 5307-10, (2012)

Loading...


Chemical analysis of late stages of pheomelanogenesis: conversion of dihydrobenzothiazine to a benzothiazole structure. Wakamatsu K, Ohtara K, and Ito S Pigment Cell Melanoma Res. 22(4), 474-86, (2009)

Loading...


Chemical degradation of melanins: application to identification of dopamine-melanin. Ito S and Wakamatsu K Pigment Cell Res. 11(2), 120-6, (1998)

Loading...


Effects of select anions from the Hofmeister series on the gas-phase conformations of protein ions measured with traveling-wave ion mobility spectrometry/mass spectrometry. Merenbloom SI, Flick TG, Daly MP, et al. J. Am. Soc. Mass Spectrom. 22(11), 1978-90, (2011)

Loading...


HPLC analysis of pheomelanin degradation products in human urine. Takasaki A, Nezirević D, Arstrand K, et al. Pigment Cell Res. 16(5), 480-6, (2003)

Loading...


Investigation of the impurities found in methamphetamine synthesised from pseudoephedrine by reduction with hydriodic acid and red phosphorus. Windahl KL, McTigue MJ, Pearson JR, et al. Forensic Sci. Int. 76(2), 97-114, (1995)

Loading...


Investigation of the reaction impurities associated with methylamphetamine synthesized using the Nagai method. Kunalan V, Kerr WJ, and Daéid NN Anal. Chem. 84(13), 5744-52, (2012)

Loading...


Hydroiodic acid attachment kinetics as a chemical probe of gaseous protein ion structure: bovine pancreatic trypsin inhibitor. Stephenson JL, Schaaff TG, and McLuckey SA J. Am. Soc. Mass Spectrom. 10(6), 552-6, (1999)

Loading...


Carbonic anhydrase inhibitors. Inhibition studies with anions and sulfonamides of a new cytosolic enzyme from the scleractinian coral Stylophora pistillata. Anthony Bertucci et al Bioorg. Med. Chem. Lett. 21, 710-4, (2011)

Loading...


Low-level bromate analysis in drinking water by ion chromatography with optimized suppressed conductivity cell current followed by a post-column reaction and UV/Vis detection. Fotsing M, Barbeau B, and Prevost M J. Environ. Sci. Health. A. Tox. Hazard. Subst. Environ. Eng. 46(4), 420-5, (2011)

Loading...


Spectrophotometric assay of eumelanin in tissue samples. Ito S, Wakamatsu K, and Ozeki H Anal. Biochem. 215(2), 273-7, (1993)

Loading...


Selective reduction of biomass by hydriodic acid and its in situ regeneration from iodine by metal/hydrogen. Yang W, Grochowski MR, and Sen A ChemSusChem 5(7), 1218-22, (2012)

Loading...


"Total" acidic metabolites of catecholamines in urine as determined by hydrolysis with hydriodic acid and liquid chromatography: application to patients with neuroblastoma and melanoma. Ito S, Kato T, Maruta K, et al. Clin. Chem. 31(7), 1185-8, (1985)

Loading...


Iodide-catalyzed reductions: development of a synthesis of phenylacetic acids. Milne JE, Storz T, Colyer JT, et al. J. Org. Chem. 76(22), 9519-24, (2011)

Loading...


Melanin biosynthesis in Cryptococcus neoformans. Williamson PR, Wakamatsu K, and Ito S J. Bacteriol. 180(6), 1570-2, (1998)

Loading...


Myoglobinuria and hypocalcemia after a superficial hydriodic acid burn. Lionelli GT, Pickus EJ, Bray JG, et al. J. Burn Care Rehabil. 22(5), 341-5, (2001)

Loading...


Photodissociation of hydrated hydrogen iodide clusters. Lee HM, Kolaski M, and Kim KS ChemPhysChem 9(4), 567-71, (2008)

Loading...


Paired hydrogen bonds in the hydrogen halide homodimer (HI)2. Wang Z, McElmurry BA, Lucchese RR, et al. J. Chem. Phys. 134(6), 064317, (2011)

Loading...


Concentrated hydriodic acid in simultaneous deprotections of multifunctional inositols. Miethchen R, Schmidt A, Neitzel K, et al. Carbohydr. Res. 340(4), 741-8, (2005)

Loading...


High-performance liquid chromatography of N-alkylprotoporphyrins: an investigation of their formation and loss under chemical and enzymatic conditions in vitro. Gibbs AH and De Matteis F Anal. Biochem. 196(2), 215-22, (1991)

Loading...


Indirect spectrophotometric determination of small amounts of selenium(IV) and arsenic(V) by simple extraction using flotation columns. Mostafa GA and Ghazy SE Anal. Sci. 17(10), 1189-93, (2001)

Loading...


Hydriodic acid to overcome tubal occlusion in sterility. GARRETT SS Am. J. Surg. 80(1), 131-2, (1950)

Loading...


Merck 14,4868 / Merck 14,4776 / Merck 14,4797

Fieser 1,449 / Fieser 2,213 / Fieser 6,281 / Fieser 7,173 / Fieser 8,246 / Fieser 9,238 / Fieser 10,199 / Fieser 15,166

Aldrich MSDS 1, 1047:A / Corp MSDS 2 (1), 1891:C / RegBook 1 (3), 3253:F / Sax 6, 1544

Corp MSDS 2 (1), 1891:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?