EMAIL THIS PAGE TO A FRIEND
21007 Sigma-Aldrich

Cesium ionophore I

Selectophore, function tested

Synonym: Calix[6]arene-hexaacetic acid hexaethylester

  • CAS Number 97600-45-8

  • Empirical Formula (Hill Notation) C66H72O18

  • Molecular Weight 1153.27

  •  Beilstein Registry Number 5231462

  •  MDL number MFCD00214147

  •  PubChem Substance ID 329752074

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   LSVIETSWDFUREK-UHFFFAOYSA-N
quality   function tested

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Cs-ionophore used in PVC membrane electrodes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

83757 Cesium ionophore II, Selectophore, function tested, ≥97.0%
01657 Cesium Ionophore III, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?