• USA Home
  • 21189 - Calcium ionophore II - membrane A

EMAIL THIS PAGE TO A FRIEND
21189 Sigma-Aldrich

Calcium ionophore II - membrane A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ready-to-use Membranes More...
grade   Selectophore

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ion-selective membrane for assaying calcium with ISE in sub-nanomolar range

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?