• USA Home
  • 21196 - Calcium ionophore II - cocktail A

EMAIL THIS PAGE TO A FRIEND
21196 Sigma-Aldrich

Calcium ionophore II - cocktail A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Calcium-selective membrane solution for microelectrodes suitable for the extra- and intracellular assay of calcium-activity

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

99371 Calcium ionophore II - cocktail A, Selectophore

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?