• USA Home
  • 21249 - (+)-Calcium L-tartrate dihydrate

EMAIL THIS PAGE TO A FRIEND
21249 Sigma-Aldrich

(+)-Calcium L-tartrate dihydrate

purum, ≥97.0% (KT)

  • CAS Number 3164-34-9

  • Linear Formula C4H4O6Ca · 2H2O

  • Molecular Weight 224.18

  •  Beilstein Registry Number 6121722

  •  EC Number 221-621-5

  •  MDL number MFCD00150687

  •  PubChem Substance ID 329752122

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Salts, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   GUPPESBEIQALOS-ZVGUSBNCSA-L
grade   purum
assay   ≥97.0% (KT)
optical activity   [α]20/D +7.3±0.3°, c = 1% in 1 M HCl

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?