• USA Home
  • 21262 - Calix[4]arene-25,26,27,28-tetrol

EMAIL THIS PAGE TO A FRIEND
21262 Sigma-Aldrich

Calix[4]arene-25,26,27,28-tetrol

purum, ≥97.0% (HPLC)

Synonym: Calix[4]arene

  • CAS Number 74568-07-3

  • Empirical Formula (Hill Notation) C28H24O4

  • Molecular Weight 424.49

  •  Beilstein Registry Number 3574428

  •  MDL number MFCD00233673

  •  PubChem Substance ID 329752125

Purchase

Properties

Related Categories Calixarenes, Chelation/Complexation Compounds, Chemical Synthesis, Synthetic Reagents
InChI Key   YPNHVQZZPXPQOS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   310-314 °C

Description

Other Notes

Starting material for preparing functionalized calix(4)arenes

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

423246 Calix[4]arene-25,26,27,28-tetrol, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?