EMAIL THIS PAGE TO A FRIEND
21282 Sigma-Aldrich

(+)-Camphanic acid

purum, ≥98.0% (T)

Synonym: (1R)-3-Oxo-4,7,7-trimethyl-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid, (1R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

  • CAS Number 67111-66-4

  • Empirical Formula (Hill Notation) C10H14O4

  • Molecular Weight 198.22

  •  Beilstein Registry Number 3609343

  •  MDL number MFCD00005535

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Lactones, Organic Building Blocks More...
InChI Key   KPWKPGFLZGMMFX-ZJUUUORDSA-N
grade   purum
assay   ≥98.0% (T)
optical activity   [α]20/D +20.0±1°, c = 1% in dioxane
mp   199-204 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

382418 (1R)-(+)-Camphanic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?