Synonym: Cuprous bromide
CAS Number 7787-70-4
Linear Formula CuBr
Molecular Weight 143.45
EC Number 232-131-6
MDL number MFCD00010969
PubChem Substance ID 24852749
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Biology, Chemical Synthesis, Click Chemistry: Catalysts, Ligands and Reagents, Conjugation Chemistry: Azides, Alkynes and Other Reagents, |
| assay | 98% |
| mp | 504 °C(lit.) |
| density | 4.71 g/mL at 25 °C(lit.) |
1 kg in glass bottle
250 g in poly bottle
50 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?