Synonym: Copper monochloride, Cuprous chloride
CAS Number 7758-89-6
Linear Formula CuCl
Molecular Weight 99.00
EC Number 231-842-9
MDL number MFCD00010971
PubChem Substance ID 24852756
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | reagent grade |
| vapor pressure | 1.3 mmHg ( 546 °C) |
| assay | 97% |
| bp | 1490 °C(lit.) |
| mp | 430 °C(lit.) |
2 kg in poly bottle
25, 500 g in poly bottle
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?