EMAIL THIS PAGE TO A FRIEND
213322 Sigma-Aldrich

Calcium hydride Green Alternative

chunks, +4 mesh, reagent grade, 95%

Purchase

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Alternative Energy, Calcium Salts, Chemical Synthesis, Enabling Products,
grade   reagent grade
InChI Key   FAQLAUHZSGTTLN-UHFFFAOYSA-N
assay   95%
form   chunks
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
particle size   +4 mesh
mp   190 °C(lit.)

Description

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1404 4.3 / PGI
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?