Synonym: Salmiac
CAS Number 12125-02-9
Linear Formula NH4Cl
Molecular Weight 53.49
Beilstein Registry Number 4371014
EC Number 235-186-4
MDL number MFCD00011420
PubChem Substance ID 24852773
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, Ammonium, Chemical Synthesis, Essential Chemicals, Inorganic Salts, |
| grade | ACS reagent |
| vapor density | 1.9 (vs air) |
| vapor pressure | 1 mmHg ( 160.4 °C) |
| assay | ≥99.5% |
| impurities | ≤0.005% insolubles |
| ign. residue | ≤0.01% |
| pH | 4.5-5.5 (25 °C, 5%) |
12 kg in poly drum
2.5, 4×2.5 kg in poly bottle
25, 500, 6×500 g in poly bottle
50 kg in fiber drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?