• USA Home
  • 21350-U - Hamilton® chaney adapter

EMAIL THIS PAGE TO A FRIEND
21350-U Sigma-Aldrich

Hamilton® chaney adapter

for use with Hamilton 700, 1000, 1700 series (except 10μL), for use with Hamilton 7000 series (0.5 - 5μL)

Purchase

Properties

Related Categories Analytical/Chromatography, Hamilton Chaney Adaptors, Syringe Accessories, Syringes
material   aluminum
packaging   pack of 1
mfr. no.   Hamilton, 14725
W × L   2.3 cm × 11 cm
compatibility   for use with Hamilton 700, 1000, 1700 series (except 10μL)
  for use with Hamilton 7000 series (0.5 - 5μL)

Description

General description

Recommended for sample reproducibility.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?