• USA Home
  • 21382 - (1R)-(−)-10-Camphorsulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
21382 Sigma-Aldrich

(1R)-(−)-10-Camphorsulfonyl chloride

for chiral derivatization, 97.0-103.0% (AT)

Synonym: (−)-Camphor-10-sulfonyl chloride, (1R)-Camphor-10-sulfonic acid chloride

  • CAS Number 39262-22-1

  • Empirical Formula (Hill Notation) C10H15ClO3S

  • Molecular Weight 250.74

  •  Beilstein Registry Number 3205975

  •  MDL number MFCD00064155

  •  PubChem Substance ID 329752151

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents HPLC, UV-VIS More...
grade   for chiral derivatization
InChI Key   BGABKEVTHIJBIW-XVKPBYJWSA-N
assay   97.0-103.0% (AT)
optical activity   [α]20/D −32±2°, c = 1% in chloroform
mp   66-68 °C(lit.)
  66-68 °C
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?