EMAIL THIS PAGE TO A FRIEND
21460 Sigma-Aldrich

Decanoyl chloride

purum, ≥98.0% (GC)

Synonym: Capric acid chloride

  • CAS Number 112-13-0

  • Linear Formula CH3(CH2)8COCl

  • Molecular Weight 190.71

  •  Beilstein Registry Number 507055

  •  EC Number 203-938-0

  •  MDL number MFCD00000771

  •  PubChem Substance ID 329752155

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor density   6.57 (vs air)
InChI Key   IPIVAXLHTVNRBS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.441(lit.)
  n20/D 1.441
bp   94-96 °C/5 mmHg(lit.)
density   0.933 g/mL at 20 °C
  0.919 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
222.8 °F
Flash Point(C) 
106 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

140295 Decanoyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?