Synonym: Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt
CAS Number 7783-85-9
Linear Formula (NH4)2Fe(SO4)2 · 6H2O
Molecular Weight 392.14
EC Number 233-151-8
MDL number MFCD00150530
PubChem Substance ID 24852891
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 98.5-101.5% |
| 99% | |
| impurities | ≤0.01% insolubles |
| ≤0.05% Not NH4OH pptd. | |
| mp | 100 °C (dec.)(lit.) |
| anion traces | phosphate (PO43-): ≤0.003% |
100, 500 g in poly bottle
12 kg in poly drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?