CAS Number 7758-05-6
Linear Formula KIO3
Molecular Weight 214.00
EC Number 231-831-9
MDL number MFCD00011406
PubChem Substance ID 24852922
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 99.4-100.4% (ACS specification) |
| 99.5% | |
| impurities | ≤0.005% N compounds |
| ≤0.005% insolubles | |
| pH | 5.0-8.0 (25 °C, 5%) |
| mp | 560 °C(lit.) |
100, 500 g in poly bottle
12 kg in poly drum
2.5, 4×2.5 kg in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?