Synonym: Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)
CAS Number 7783-28-0
Linear Formula (NH4)2HPO4
Molecular Weight 132.06
EC Number 231-987-8
MDL number MFCD00010891
PubChem Substance ID 24852928
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Ammonium, Chemical Synthesis, Essential Chemicals, Inorganic Salts, |
| grade | ACS reagent |
| assay | ≥98% |
| impurities | ≤0.005% NH4OH pptd. |
| ≤0.005% insolubles | |
| pH | 7.7-8.1 (25 °C, 5%) |
| mp | 155 °C (dec.)(lit.) |
| anion traces | chloride (Cl-): ≤0.001% |
100, 500 g in poly bottle
12 kg in fiber drum
2.5 kg in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?