Synonym: Potassium peroxodisulfate
CAS Number 7727-21-1
Linear Formula K2S2O8
Molecular Weight 270.32
EC Number 231-781-8
MDL number MFCD00011386
PubChem Substance ID 24852943
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Free Radical Initiators, Inorganic Peroxides, |
| grade | ACS reagent |
| vapor density | 9.3 (vs air) |
| assay | ≥99.0% |
| impurities | ≤0.001% Cl compounds |
| ≤0.005% insolubles | |
| cation traces | Fe: ≤5 ppm |
| Mn: ≤2 ppm |
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?