EMAIL THIS PAGE TO A FRIEND
216232 Sigma-Aldrich

Sodium persulfate

reagent grade, ≥98%

Synonym: Sodium peroxodisulfate

Purchase

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

2.5 kg in poly bottle

25, 500 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1505 5.1 / PGIII
WGK Germany 
1
RTECS 
SE0525000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?