Synonym: Cuprous cyanide
CAS Number 544-92-3
Linear Formula CuCN
Molecular Weight 89.56
Beilstein Registry Number 3587244
EC Number 208-883-6
MDL number MFCD00010975
PubChem Substance ID 24852948
Popular Documents: Specification Sheet (PDF)
| assay | 99% |
| mp | 474 °C(lit.) |
| density | 2.92 g/mL at 25 °C(lit.) |
Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.1
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?