CAS Number 7722-84-1
Empirical Formula (Hill Notation) H2O2
Molecular Weight 34.01
Beilstein Registry Number 3587191
MDL number MFCD00011333
PubChem Substance ID 24852978
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, Acids & Bases, Bioactive Small Molecules, Blocking Sera and Reagents, Cell Biology, |
| grade | ACS reagent |
| vapor density | 1 (vs air) |
| vapor pressure | 23.3 mmHg ( 30 °C) |
| assay | 29.0-32.0% (ACS specification) |
| quality | stabilized |
| concentration | 30 wt. % in H2O |
| impurities | ≤0.0006 meq/g Titr. acid |
Live Chat and Frequently Asked Questions
are available for this Product.
100, 500 mL in poly bottle
4 L in poly bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?