EMAIL THIS PAGE TO A FRIEND
21700 Sigma-Aldrich

Octanoyl chloride

purum, ≥99.0% (GC)

Synonym: Capryloyl chloride

  • CAS Number 111-64-8

  • Linear Formula CH3(CH2)6COCl

  • Molecular Weight 162.66

  •  Beilstein Registry Number 635917

  •  EC Number 203-891-6

  •  MDL number MFCD00000766

Purchase

Properties

vapor pressure   2.5 mmHg ( 20 °C)
InChI Key   REEZZSHJLXOIHL-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.435(lit.)
  n20/D 1.435
bp   195 °C(lit.)
  77 °C/11 mmHg(lit.)
density   0.949 g/mL at 20 °C
  0.953 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3265 8/PG 2
WGK Germany 
3
RTECS 
RH1570000
Flash Point(F) 
180 °F
Flash Point(C) 
82 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

O4733 Octanoyl chloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?