• USA Home
  • 21920 - O-(Carboxymethyl)hydroxylamine hemihydrochloride

EMAIL THIS PAGE TO A FRIEND
21920 Sigma-Aldrich

O-(Carboxymethyl)hydroxylamine hemihydrochloride

purum, ≥97.0% (AT)

Synonym: (Aminooxy)acetic acid hemihydrochloride, (Carboxymethoxy)amine hemihydrochloride, Hydroxylamine-O-acetic acid hemihydrochloride

  • CAS Number 2921-14-4

  • Linear Formula NH2OCH2COOH · 0.5HCl

  • Molecular Weight 109.30

  •  Beilstein Registry Number 3680528

  •  EC Number 220-862-3

  •  MDL number MFCD00012955

  •  PubChem Substance ID 24853112

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydroxylamines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   KBXIJIPYZKPDRU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   156 °C (dec.)(lit.)

Description

Other Notes

Reagent for introducing an acid group in ketones; used e.g. to prepare chemiluminescent ABEI labels from ketones

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
AF3150000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?