• USA Home
  • 21949 - 6-(4-Carboxyphenyl)-3,8-diamino-5-methylphenanthridinium chloride

EMAIL THIS PAGE TO A FRIEND
21949 Sigma-Aldrich

6-(4-Carboxyphenyl)-3,8-diamino-5-methylphenanthridinium chloride

≥87.0% (HPLC)

Synonym: 4′-Carboxydimidium chloride

  • CAS Number 52671-19-9

  • Empirical Formula (Hill Notation) C21H18ClN3O2

  • Molecular Weight 379.84

  •  Beilstein Registry Number 4085139

  •  MDL number MFCD00214263

  •  PubChem Substance ID 329752225

Purchase

Properties

InChI Key   BDPDBMJZOJIFLS-UHFFFAOYSA-N
assay   ≥87.0% (HPLC)
storage temp.   −20°C

Description

Other Notes

DNA intercalator. In the synthesis of a DNA capture reagent

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?