• USA Home
  • 22020 - (±)-Carnitine hydrochloride

EMAIL THIS PAGE TO A FRIEND
22020 Sigma-Aldrich

(±)-Carnitine hydrochloride

purum, ≥99.0% (AT)

Synonym: β-Hydroxy-γ-(trimethylammonio)butyrate hydrochloride, (±)-Carnitine chloride, DL-Carnitine hydrochloride

  • CAS Number 461-05-2

  • Linear Formula (CH3)3N(Cl)CH2CH(OH)CH2COOH

  • Molecular Weight 197.66

  •  Beilstein Registry Number 4163212

  •  EC Number 207-309-1

  •  MDL number MFCD00011904

  •  PubChem Substance ID 329752235

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   JXXCENBLGFBQJM-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (AT)
mp   197-199 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BP2979140

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

C9500 (±)-Carnitine hydrochloride, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?