Synonym: Mercuric oxide yellow
CAS Number 21908-53-2
Linear Formula HgO
Molecular Weight 216.59
EC Number 244-654-7
MDL number MFCD00011045
PubChem Substance ID 24853210
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | ≥99.0% |
| impurities | ≤0.005% N compounds |
| ≤0.03% insol. dil. HCl | |
| reduction residue | ≤0.05% |
| mp | 500 °C (dec.)(lit.) |
| anion traces | chloride (Cl-): ≤0.025% |
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?