Synonym: Ammonia aqueous, Ammonia water
CAS Number 1336-21-6
Linear Formula NH4OH
Molecular Weight 35.05
Beilstein Registry Number 3587154
MDL number MFCD00066650
PubChem Substance ID 24853221
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| Quality Level | PREMIUM |
| grade | ACS reagent |
| assay | 28.0-30.0% NH3 basis |
| autoignition temp. | 1204 °F |
| expl. lim. | 27 % |
| impurities | MnO4- reducers, passes test |
| ≤0.002% CO2 |
ACS specifications same for product 320145
1 L in glass bottle
2.5, 6×2.5 L in poly bottle
25, 100, 12×100 mL in glass bottle
View returnable container options.
174 L in poly drum
500 mL in poly bottle
6×500 mL in poly bottle
6×500 mL-GLA in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?