Synonym: Diatomaceous earth, flux-calcined
CAS Number 68855-54-9
MDL number MFCD00132803
PubChem Substance ID 24853234
| Related Categories | Adsorbents, Filter Aids and Drying Agents, Diatomaceous Earth, Essential Chemicals, Research Essentials |
| quality | flux calcined |
| treated with sodium carbonate | |
| composition | Al2O3, 4.0% |
| CaO, 0.5% | |
| Fe2O3, 1.3% | |
| Na2O + K2O, 3.3% | |
| SiO2, 89.6% |
Not Intended for cGMP Parenteral Pharmaceutical Manufacturing
1 kg in poly bottle
5 kg in fiber drum
Celite is a registered trademark of Imerys Minerals California, Inc.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?