• USA Home
  • 22160 - D-(+)-Cellobiose octaacetate

EMAIL THIS PAGE TO A FRIEND
22160 Sigma-Aldrich

D-(+)-Cellobiose octaacetate

for microbiology, ≥96.0%

  • CAS Number 5346-90-7

  • Empirical Formula (Hill Notation) C28H38O19

  • Molecular Weight 678.59

  •  Beilstein Registry Number 79280

  •  EC Number 226-304-5

  •  MDL number MFCD00009600

  •  PubChem Substance ID 329752277

Purchase

Properties

Related Categories Base Ingredients for Media, Carbohydrate Sources (Sugars/Extracts), Microbiology, Sugars for Media
grade   for microbiology
InChI Key   WOTQVEKSRLZRSX-AXXYRMHMSA-N
assay   ≥96.0% (TLC)
  ≥96.0%
optical activity   [α]20/D +41.5±2°, c = 1% in chloroform
mp   225-232 °C
solubility   chloroform: soluble0.1 g/mL, clear to almost clear, colorless to faintly brownish-yellow

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?