EMAIL THIS PAGE TO A FRIEND
22181 Sigma-Aldrich

Cellulose

from spruce, for column chromatography

Synonym: Cellulose powder, Cotton linters

Purchase

Properties

biological source   from spruce
grade   for column chromatography
fiber L   0.02-0.15 mm

Description

Application

High purity cellulose powders for partition chromatography.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
FJ5691460

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

22182 Cellulose, acid washed, from spruce, for column chromatography

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?