CAS Number 7782-68-5
Empirical Formula (Hill Notation) HIO3
Molecular Weight 175.91
EC Number 231-962-1
MDL number MFCD00011348
PubChem Substance ID 24853264
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| assay | ≥99.5% |
| impurities | ≤0.01% insolubles |
| ≤0.1% N compounds | |
| ign. residue | ≤0.02% |
| anion traces | Cl- and Br-: ≤0.02% |
| iodide (I-): ≤0.01% |
50, 250 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?