Synonym: TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate
CAS Number 10101-89-0
Linear Formula Na3PO4 · 12H2O
Molecular Weight 380.12
EC Number 231-509-8
MDL number MFCD00149198
PubChem Substance ID 24853305
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Buffers, Biological Buffers, Buffers A to Z, Chemical Synthesis, |
| grade | ACS reagent |
| assay | ≥98% |
| 98.0-102.0% (ACS specification) | |
| impurities | ≤0.01% insolubles |
| density | 1.62 g/mL at 25 °C |
| anion traces | chloride (Cl-): ≤0.001% |
| sulfate (SO42-): ≤0.01% |
2.5 kg in poly bottle
500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?