• USA Home
  • 222283 - Magnesium perchlorate

EMAIL THIS PAGE TO A FRIEND

222283 Sigma-Aldrich

Magnesium perchlorate

ACS reagent

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Magnesium Salts, Oxidizing Agents, Perchlorates,
grade   ACS reagent
impurities   ≤0.005 meq/g Titr. free acid
  ≤0.025 meq/g Titr. base
loss   ≤8% loss on drying
suitability   passes test for moisture absorption

Description

Packaging

100 g in poly bottle

5 g in glass bottle

500 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-high Purity Salts

Customers Also Viewed

Ascarite<SUP>®</SUP>

Sodium hydroxide-coated silica, 8-20 mesh

Ascarite<SUP>®</SUP>

Sodium hydroxide-coated silica, 20-30 mesh

Magnesium perchlorate

puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)

Magnesium perchlorate

puriss. p.a., drying agent, ACS reagent, ≥98.0% (calc. based on dry substance, KT)

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
7/8-17-26-27-36/37/39
RIDADR 
UN 1475 5.1 / PGII
WGK Germany 
1
RTECS 
SC8925000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Synthesis and photoinduced electron transfer of phthalocyanine-perylenebisimide pentameric arrays. Céspedes-Guirao FJ, Ohkubo K, Fukuzumi S, et al. J. Org. Chem. 74(16), 5871-80, (2009)

Loading...


[The experimental effect of growth stimulants on protein assimilation]. Nikolaevskaia VR and Volkova LD Vopr. Pitan. (3), 58-60, (1990)

Loading...


Magnesium perchlorate and cis-platinum as radiosensitizers in in vitro radiolysis of nucleotides. Anastassopoulou JD Magnes. Res. 3(1), 19-21, (1990)

Loading...


The dose rate effects on the in vitro radiolysis products of magnesium-guanosine-5'-monophosphate complexes in aqueous solutions. Anastassopoulou JD, Rakintzis NT, and Theophanides T Magnes. Res. 3(1), 15-8, (1990)

Loading...


Chloranil-photosensitized monomerization of dimethylthymine cyclobutane dimers and effect of magnesium perchlorate. Pac C, Miyamoto I, Masaki Y, et al. Photochem. Photobiol. 52(5), 973-9, (1990)

Loading...


Ligand exchange dynamics in aqueous solution studied with 2DIR spectroscopy. Park S, Ji M, and Gaffney KJ J. Phys. Chem. B 114(19), 6693-702, (2010)

Loading...


Flavin-photosensitized monomerization of dimethylthymine cyclobutane dimer in the presence of magnesium perchlorate. Miyake K, Masaki Y, Miyamoto I, et al. Photochem. Photobiol. 58(5), 631-6, (1993)

Loading...


Notable effects of the metal salts on the formation and decay reactions of α-tocopheroxyl radical in acetonitrile solution. The complex formation between α-tocopheroxyl and metal cations. Kohno Y, Fujii M, Matsuoka C, et al. J. Phys. Chem. B 115(32), 9880-8, (2011)

Loading...


Ultrafast two-dimensional infrared spectroscopy resolves the conformational change of an Evans auxiliary induced by Mg(ClO4)2. Messmer AT, Steinwand S, Lippert KM, et al. J. Org. Chem. 77(24), 11091-5, (2012)

Loading...


Transformation of D-(-)-ribose into a natural product-like scaffold via a Lewis acid catalyzed intramolecular hetero-Diels-Alder reaction. Messer R, Fuhrer CA, and Häner R Nucleosides Nucleotides Nucleic Acids 26(6-7), 701-4, (2007)

Loading...


Biological implications of magnesium salts at the molecular level. Theophanides T Magnes. Res. 9(4), 259-62, (1996)

Loading...


Merck 14,5678

Corp MSDS 1 (2), 2180:D / FT-IR 1 (2), 1254:A / FT-IR 2 (3), 4762:A / RegBook 1 (3), 3457:C / Sax 6, 2144

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?