Synonym: Calcined soda, Carbonic acid disodium salt, Soda ash
CAS Number 497-19-8
Linear Formula Na2CO3
Molecular Weight 105.99
Beilstein Registry Number 4154566
EC Number 207-838-8
MDL number MFCD00003494
PubChem Substance ID 24853386
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Bases, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent (primary standard) |
| anhydrous | |
| assay | 99.95-100.05% dry basis |
| impurities | ≤0.005% silica |
| ≤0.01% insolubles | |
| loss | ≤1.0% loss on heating |
| mp | 851 °C(lit.) |
Buffer component, may be used for the removal of peripheral membrane proteins.
500, 6×500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?