224111 Sigma-Aldrich

Ethylene glycol diethyl ether

98%

DOWNLOAD MSDS (PDF)

Synonym: 1,2-Diethoxyethane

  • CAS Number 629-14-1

  • Linear Formula CH3CH2OCH2CH2OCH2CH3

  • Molecular Weight 118.17

  •  Beilstein Registry Number 1732917

  •  EC Number 211-076-1

  •  MDL number MFCD00009253

  •  PubChem Substance ID 24853420

Properties

Related Categories ACS and Reagent Grade Solvents, Amber Glass Bottles, Reagent, Reagent Grade Solvents, Solvent Bottles,
vapor pressure   9.4 mmHg ( 20 °C)
assay   98%
refractive index   n20/D 1.3923(lit.)
bp   121 °C(lit.)
mp   −74 °C(lit.)
density   0.842 g/mL at 25 °C(lit.)

Description

Packaging

5, 100, 500 mL in glass bottle

Price and Availability

Customers Also Viewed

1,2-Dimethoxyethane

anhydrous, 99.5%

Diethylene glycol diethyl ether

reagent grade, ≥98%

Diethylene glycol dimethyl ether

anhydrous, 99.5%

Ethylene glycol

anhydrous, 99.8%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?