Synonym: Ammonium bifluoride, Etching powder
CAS Number 1341-49-7
Linear Formula NH4HF2
Molecular Weight 57.04
EC Number 215-676-4
MDL number MFCD00012018
PubChem Substance ID 24853457
Popular Documents: Specification Sheet (PDF)
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, Research Essentials |
| grade | reagent grade |
| assay | 95% |
| mp | 125 °C(lit.) |
2.5 kg in poly bottle
25, 500 g in poly bottle
Employed in a new hydrothermal synthesis of complex flourides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important forsolid-state lasers as well as for photoluminescence and magnetic characteristics.1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?