EMAIL THIS PAGE TO A FRIEND
22500 Sigma-Aldrich

Chelidonic acid

purum, anhydrous, ≥98.0% (T)

Synonym: 4-Oxo-4H-pyran-2,6-dicarboxylic acid

  • CAS Number 99-32-1

  • Empirical Formula (Hill Notation) C7H4O6

  • Molecular Weight 184.10

  •  Beilstein Registry Number 163607

  •  EC Number 202-749-0

  •  MDL number MFCD00006577

  •  PubChem Substance ID 329752321

Purchase

Properties

Related Categories Berberis sp., Building Blocks, Cell Biology, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   PBAYDYUZOSNJGU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
quality   anhydrous
ign. residue   ≤0.5%
mp   265 °C (dec.)(lit.)
solubility   hot water: soluble0.1g in 10mL, clear (colorless to faintly red or slightly pink)

Description

Application

Chelidonic acid was used as an inhibitor of rat brain glutamate decarboxylase.

General description

Chelidonic acid (a γ-pyrone) has multiple pharmacological effects like mild analgesic, an oncostatic, an antimicrobial and a central nervous system sedative.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UP7350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?