EMAIL THIS PAGE TO A FRIEND
22590 Sigma-Aldrich

Quinhydrone

puriss.

Synonym: Hydroquinone: benzoquinone 1:1 complex

  • CAS Number 106-34-3

  • Linear Formula C6H4(OH)2 · C6H4O2

  • Molecular Weight 218.21

  •  Beilstein Registry Number 3919222

  •  EC Number 203-387-6

  •  MDL number MFCD00010310

  •  PubChem Substance ID 329752328

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   BDJXVNRFAQSMAA-UHFFFAOYSA-N
grade   puriss.
composition   benzoquinone, ~50% HPLC
  hydroquinone, ~50% HPLC
mp   167-172 °C(lit.)
  167-172 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
VA4550000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

282960 Quinhydrone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?